Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207469.2 | NP_997352.1 | 2 | 181 | ||
GenBank mRNA | AF525932.1 | - | 2 | 138 | ||
AY358708.1 | - | 2 | 181 | |||
AY621329.1 | - | 2 | 138 | |||
BC137292.1 | - | 2 | 175 | 9021289 | ||
BC137293.1 | - | 2 | 175 | 9021290 | ||
EF426755.1 | - | 2 | 181 |