Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNB4 SIRNA Assays ›
Gene Symbol:
CACNB4
Gene Name:
calcium voltage-gated channel auxiliary subunit beta 4
Gene Aliases:
CAB4, CACNLB4, EA5, EIG9, EJM, EJM4, EJM6
Chromosome Location:
Chr.2: 151832771 - 152099988 on Build GRCh38
UniGene ID:
Hs.120725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533854
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNB4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000726.3
NP_000717.2
4
426
NM_001005746.2
NP_001005746.1
4
376
NM_001005747.2
NP_001005747.1
3
514
NM_001145798.1
NP_001139270.1
4
426
NM_001320722.1
NP_001307651.1
4
674
XM_011511795.2
XP_011510097.1
4
2143
XM_011511796.2
XP_011510098.1
3
1097
XM_011511797.2
XP_011510099.1
4
1335
XM_011511798.2
XP_011510100.1
4
1335
XM_017004880.1
XP_016860369.1
4
427
XM_017004881.1
XP_016860370.1
3
495
XM_017004882.1
XP_016860371.1
4
1820
XM_017004883.1
XP_016860372.1
5
1965
XM_017004884.1
XP_016860373.1
4
1819
XM_017004885.1
XP_016860374.1
3
1659
XM_017004887.1
XP_016860376.1
4
1335
XM_017004888.1
XP_016860377.1
4
435
GenBank mRNA
AB302276.1
-
4
370
AF038852.1
-
4
336
AK290049.1
-
3
495
AK291378.1
-
5
509
AK294398.1
-
4
350
AK316045.1
-
4
367
AY054985.1
-
3
485
BC075049.2
-
4
377
30915578
BP229182.1
-
3
482
BP360802.1
-
4
359
BX279713.1
-
4
376
BX648679.1
-
3
301
HQ214056.1
-
3
256
U95020.1
-
4
426
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
transport
cellular calcium ion homeostasis
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
neuromuscular junction development
adult walking behavior
gamma-aminobutyric acid secretion
neuronal action potential propagation
synaptic transmission, glutamatergic
cAMP metabolic process
spleen development
thymus development
Peyer's patch development
muscle fiber development
T cell receptor signaling pathway
detection of light stimulus involved in visual perception
membrane depolarization
cardiac conduction
calcium ion transmembrane transport
regulation of voltage-gated calcium channel activity
voltage-gated calcium channel activity
calcium channel activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0