Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000275.2 | NP_000266.2 | 4 | 524 | ||
NM_001300984.1 | NP_001287913.1 | 4 | 524 | |||
XM_011521640.2 | XP_011519942.1 | 4 | 554 | |||
XM_017022255.1 | XP_016877744.1 | 4 | 505 | |||
XM_017022256.1 | XP_016877745.1 | 4 | 1002 | |||
XM_017022257.1 | XP_016877746.1 | 4 | 527 | |||
XM_017022258.1 | XP_016877747.1 | 4 | 527 | |||
XM_017022259.1 | XP_016877748.1 | 4 | 527 | |||
XM_017022260.1 | XP_016877749.1 | 4 | 527 | |||
XM_017022261.1 | XP_016877750.1 | 3 | 1021 | |||
XM_017022262.1 | XP_016877751.1 | 4 | 527 | |||
XM_017022263.1 | XP_016877752.1 | 4 | 527 | |||
XM_017022264.1 | XP_016877753.1 | 4 | 527 | |||
XM_017022265.1 | XP_016877754.1 | 4 | 527 | |||
GenBank mRNA | BC012097.1 | - | 4 | 524 | 4641135 | |
M99564.1 | - | 4 | 466 |