Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005732.3 | NP_005723.2 | 6 | 1234 | ||
GenBank mRNA | AF057299.1 | - | 6 | 1154 | ||
AF057300.1 | - | 6 | 1089 | |||
AK290597.1 | - | 6 | 1176 | |||
AK307439.1 | - | 6 | 909 | |||
BC062603.1 | - | 6 | 1151 | |||
BC073850.1 | - | 6 | 1151 | |||
BC108282.1 | - | 6 | 1171 | |||
BC136436.1 | - | 6 | 1126 | 9020425 | ||
U63139.1 | - | 6 | 1221 |