Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244644.1 | NP_001231573.1 | 2 | 412 | ||
NM_014043.3 | NP_054762.2 | 3 | 504 | |||
XM_011533576.2 | XP_011531878.1 | 3 | 335 | |||
GenBank mRNA | AF151842.1 | - | 3 | 463 | ||
AK002165.1 | - | 3 | 477 | |||
AK002180.1 | - | 3 | 482 | |||
AK222654.1 | - | 3 | 464 | |||
AK296072.1 | - | 2 | 404 | |||
AK316585.1 | - | 3 | 468 | |||
AL080122.1 | - | 3 | 477 | |||
BC001553.1 | - | 3 | 320 | 3460712 | ||
BI821848.1 | - | 3 | 481 | |||
BM828410.1 | - | 3 | 504 | |||
CR533456.1 | - | 3 | 244 | |||
DA426159.1 | - | 3 | 475 |