Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004750.4 | NP_004741.1 | 7 | 1334 | ||
XM_011528422.2 | XP_011526724.1 | 7 | 1416 | |||
XM_011528423.2 | XP_011526725.1 | 7 | 1140 | |||
XM_011528424.2 | XP_011526726.1 | 7 | 1408 | |||
GenBank mRNA | AF059293.1 | - | 7 | 1258 | ||
AF073515.1 | - | 7 | 1343 | |||
AF178684.1 | - | 7 | 1191 | |||
AY358291.1 | - | 7 | 1270 |