Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302652.1 | NP_001289581.1 | 3 | 393 | ||
NM_001302653.1 | NP_001289582.1 | 3 | 393 | |||
NM_001302654.1 | NP_001289583.1 | 3 | 393 | |||
NM_032374.4 | NP_115750.2 | 3 | 393 | |||
NR_126431.1 | 4 | 558 | ||||
GenBank mRNA | AK223092.1 | - | 3 | 364 | ||
AK223103.1 | - | 3 | 364 | |||
BC007412.2 | - | 3 | 356 | 2989951 | ||
BF528062.1 | - | 4 | 460 | |||
BP317962.2 | - | 3 | 389 | |||
BP368962.1 | - | 3 | 387 | |||
BU181154.1 | - | 388 | ||||
BX119699.1 | - | 3 | 273 | |||
CB136383.1 | - | 3 | 388 | |||
CB993873.1 | - | 3 | 390 |