Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289807.1 | NP_001276736.1 | 3 | 571 | ||
NM_001289808.1 | NP_001276737.1 | 2 | 600 | |||
NM_001885.2 | NP_001876.1 | 3 | 605 | |||
XM_011542608.1 | XP_011540910.1 | 3 | 586 | |||
XM_011542609.2 | XP_011540911.1 | 2 | 827 | |||
GenBank mRNA | AF007162.1 | - | 2 | 327 | ||
AK295498.1 | - | 2 | 390 | |||
AK314029.1 | - | 2 | 343 | |||
BC007008.1 | - | 2 | 344 | 3933748 | ||
BI759672.1 | - | 3 | 515 | |||
BM721803.1 | - | 3 | 566 | |||
BT006770.1 | - | 2 | 298 | |||
FJ224314.1 | - | 2 | 323 | |||
FJ876064.1 | - | 2 | 298 | |||
KM220603.1 | - | 2 | 312 | |||
KM220604.1 | - | 2 | 312 | |||
KM220605.1 | - | 2 | 312 | |||
KM220606.1 | - | 2 | 322 | |||
KM220607.1 | - | 2 | 277 | |||
S45630.1 | - | 2 | 323 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK002135.1 | - | 1 | 542 | Chr. 11 - 111908926 - 111910893 [-] on Build GRCh38 |