Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 14 | 2598 | ||
NR_027299.1 | 13 | 2474 | ||||
GenBank mRNA | AK222844.1 | - | 13 | 2291 | ||
AK289761.1 | - | 14 | 2527 | |||
AK295711.1 | - | 14 | 2416 | |||
AK311039.1 | - | 13 | 2404 | |||
BC016620.1 | - | 14 | 2509 | 4509957 | ||
D21089.1 | - | 14 | 2599 | |||
HM113488.1 | - | 14 | 2498 | |||
KJ534962.1 | - | 14 | 2494 | |||
KJ535085.1 | - | 12 | 2210 | |||
X65024.1 | - | 13 | 2334 |