Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 11 | 1214 | ||
NM_001145809.1 | NP_001139281.1 | 11 | 1214 | |||
NM_024729.3 | NP_079005.3 | 10 | 1190 | |||
XM_006723386.3 | XP_006723449.1 | 11 | 1275 | |||
XM_011527320.2 | XP_011525622.1 | 11 | 1292 | |||
XM_011527321.2 | XP_011525623.1 | 10 | 1268 | |||
XM_011527323.2 | XP_011525625.1 | 11 | 1282 | |||
GenBank mRNA | AB290169.1 | - | 10 | 1184 | ||
AK125082.1 | - | 11 | 1293 | |||
AK307783.1 | - | 11 | 1197 | |||
AY165122.1 | - | 10 | 1190 | |||
BC018933.2 | - | 9 | 942 | |||
BC129803.1 | - | 11 | 1068 |