Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 30 | 4018 | ||
NM_001145809.1 | NP_001139281.1 | 31 | 4117 | |||
NM_024729.3 | NP_079005.3 | 29 | 3994 | |||
XM_006723386.3 | XP_006723449.1 | 30 | 4079 | |||
XM_011527320.2 | XP_011525622.1 | 30 | 4096 | |||
XM_011527321.2 | XP_011525623.1 | 29 | 4072 | |||
XM_011527323.2 | XP_011525625.1 | 30 | 4086 | |||
GenBank mRNA | AB111886.1 | - | 20 | 2676 | ||
AB290169.1 | - | 31 | 4087 | |||
AY165122.1 | - | 30 | 3994 | |||
AY203926.1 | - | 8 | 1059 | |||
BC004396.1 | - | 2 | 61 | |||
BC129803.1 | - | 18 | 2091 |