Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291006.1 | NP_001277935.1 | 3 | 346 | ||
NR_027006.1 | 3 | 346 | ||||
NR_110991.1 | 3 | 346 | ||||
XM_011530173.2 | XP_011528475.1 | 3 | 325 | |||
XM_011530174.1 | XP_011528476.1 | 3 | 325 | |||
XM_011530175.1 | XP_011528477.1 | 3 | 325 | |||
XM_011530176.2 | XP_011528478.1 | 3 | 325 | |||
XM_011530177.1 | XP_011528479.1 | 3 | 325 | |||
XM_017028797.1 | XP_016884286.1 | 3 | 325 | |||
GenBank mRNA | AK128646.1 | - | 3 | 325 | ||
BC140948.1 | - | 3 | 344 | |||
BC140950.1 | - | 3 | 344 |