Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166278.1 | NP_001159750.1 | 6, 7 | 673 | ||
NM_020730.2 | NP_065781.1 | 8, 9 | 1216 | |||
XM_005262248.3 | XP_005262305.1 | 8, 9 | 813 | |||
XM_006724625.2 | XP_006724688.1 | 14, 15 | 2262 | |||
XM_006724626.2 | XP_006724689.1 | 14, 15 | 2208 | |||
XM_017029323.1 | XP_016884812.1 | 8, 9 | 1151 | |||
XM_017029325.1 | XP_016884814.1 | 6, 7 | 2523 | |||
XM_017029326.1 | XP_016884815.1 | 6, 7 | 602 | |||
GenBank mRNA | AB033058.1 | - | 8, 9 | 1214 | ||
AK303377.1 | - | 6, 7 | 673 | |||
AK304020.1 | - | 8, 9 | 929 | |||
AK316518.1 | - | 8, 9 | 915 |