Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 13 | 1520 | ||
NM_020779.3 | NP_065830.2 | 12 | 1487 | |||
XM_011533007.2 | XP_011531309.1 | 2 | 212 | |||
GenBank mRNA | AB037757.2 | - | 13 | 1528 | ||
AK091205.1 | - | 6 | 855 | |||
AK122917.1 | - | 12 | 1412 | |||
AK315094.1 | - | 12 | 1412 | |||
AL832903.1 | - | 1 | 17 | |||
BC036659.2 | - | 12 | 1486 | 5266940 |