Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000195.4 | NP_000186.2 | 17, 18 | 2008 | ||
NM_001311345.1 | NP_001298274.1 | 16, 17 | 1961 | |||
NM_001322476.1 | NP_001309405.1 | 17, 18 | 1937 | |||
NM_001322477.1 | NP_001309406.1 | 17, 18 | 2017 | |||
NM_001322478.1 | NP_001309407.1 | 16, 17 | 1838 | |||
NM_001322479.1 | NP_001309408.1 | 16, 17 | 1918 | |||
NM_001322480.1 | NP_001309409.1 | 15, 16 | 1676 | |||
NM_001322481.1 | NP_001309410.1 | 15, 16 | 1756 | |||
NM_001322482.1 | NP_001309411.1 | 14, 15 | 1657 | |||
NM_001322483.1 | NP_001309412.1 | 16, 17 | 1794 | |||
NM_001322484.1 | NP_001309413.1 | 16, 17 | 1874 | |||
NM_001322485.1 | NP_001309414.1 | 15, 16 | 1766 | |||
NM_001322487.1 | NP_001309416.1 | 17, 18 | 2060 | |||
NM_001322489.1 | NP_001309418.1 | 15, 16 | 1818 | |||
XM_005269757.4 | XP_005269814.1 | 17, 18 | 1950 | |||
XM_017016170.1 | XP_016871659.1 | 14, 15 | 1564 | |||
XM_017016171.1 | XP_016871660.1 | 15, 16 | 1771 | |||
XM_017016172.1 | XP_016871661.1 | 14, 15 | 1582 | |||
GenBank mRNA | AL833734.1 | - | 6, 7 | 660 | ||
U65676.1 | - | 17, 18 | 1952 |