Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002473.5 | NP_002464.1 | 26, 27 | 3773 | ||
XM_011530197.2 | XP_011528499.1 | 27, 28 | 3628 | |||
XM_017028803.1 | XP_016884292.1 | 26, 27 | 3528 | |||
XM_017028804.1 | XP_016884293.1 | 26, 27 | 3528 | |||
XM_017028805.1 | XP_016884294.1 | 26, 27 | 3528 | |||
XM_017028806.1 | XP_016884295.1 | 26, 27 | 3528 | |||
GenBank mRNA | AB191263.1 | - | 26, 27 | 3673 | ||
AB290175.1 | - | 26, 27 | 3566 | |||
AK131080.1 | - | 14, 15 | 1778 | |||
BC049849.1 | - | 26, 27 | 3642 | |||
CR456526.1 | - | 26, 27 | 3574 | |||
KU159427.1 | - | 19, 20 | 3715 | |||
M31013.1 | - | 10, 11 | 1352 | |||
M81105.1 | - | 26, 27 | 3632 |