Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252835.1 | XP_005252892.1 | 6 | 1217 | ||
XM_011519956.1 | XP_011518258.1 | 6 | 2009 | |||
XM_011519957.1 | XP_011518259.1 | 6 | 1838 | |||
XM_017017428.1 | XP_016872917.1 | 6 | 1063 | |||
XM_017017429.1 | XP_016872918.1 | 6 | 1237 | |||
XM_017017431.1 | XP_016872920.1 | 6 | 962 | |||
GenBank mRNA | AK074992.1 | - | 4 | 2003 | ||
AK301558.1 | - | 5 | 1017 | |||
AK301979.1 | - | 6 | 1205 |