Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166107.1 | NP_001159579.1 | 4 | 842 | ||
NM_005518.3 | NP_005509.1 | 5 | 968 | |||
XM_011541313.1 | XP_011539615.1 | 4 | 996 | |||
GenBank mRNA | AK301594.1 | - | 4 | 625 | ||
AK302276.1 | - | 4 | 811 | |||
AK303777.1 | - | 4 | 815 | |||
AK313590.1 | - | 5 | 927 | |||
BC044217.1 | - | 5 | 921 | 5744956 | ||
CR456850.1 | - | 6 | 880 | |||
GU433940.1 | - | 3 | 445 | |||
U12789.1 | - | 4 | 447 | |||
X83618.1 | - | 5 | 931 |