Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166107.1 | NP_001159579.1 | 5 | 1017 | ||
NM_005518.3 | NP_005509.1 | 6 | 1143 | |||
XM_011541313.1 | XP_011539615.1 | 5 | 1171 | |||
GenBank mRNA | AK301594.1 | - | 5 | 800 | ||
AK302276.1 | - | 5 | 986 | |||
AK303777.1 | - | 5 | 990 | |||
AK313590.1 | - | 6 | 1102 | |||
BC044217.1 | - | 6 | 1096 | 5744956 | ||
CR456850.1 | - | 7 | 1055 | |||
U12789.1 | - | 5 | 622 | |||
X83618.1 | - | 6 | 1106 |