Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206524.1 | NP_001193453.1 | 2 | 705 | ||
NM_001206542.1 | NP_001193471.1 | 2 | 705 | |||
NM_001230.4 | NP_001221.2 | 2 | 705 | |||
NM_001306083.1 | NP_001293012.1 | 2 | 722 | |||
NM_032974.4 | NP_116756.2 | 2 | 705 | |||
NM_032976.3 | NP_116758.1 | 2 | 705 | |||
NM_032977.3 | NP_116759.2 | 2 | 705 | |||
XM_005246907.2 | XP_005246964.1 | 2 | 491 | |||
XM_006712796.3 | XP_006712859.1 | 2 | 718 | |||
XM_011511990.2 | XP_011510292.1 | 2 | 491 | |||
GenBank mRNA | AF111344.1 | - | 1 | 287 | ||
AF111345.1 | - | 1 | 287 | |||
AJ487678.1 | - | 2 | 411 | |||
AJ487679.1 | - | 2 | 408 | |||
AK225475.1 | - | 2 | 443 | |||
AK304859.1 | - | 2 | 462 | |||
AK310746.1 | - | 2 | 491 | |||
AY690601.3 | - | 2 | 443 | |||
BC042844.1 | - | 2 | 441 | 5176698 | ||
KU177959.1 | - | 1 | 287 | |||
KU177960.1 | - | 1 | 287 | |||
U60519.1 | - | 2 | 434 | |||
U86214.1 | - | 2 | 481 |