Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017423.1 | NP_001017423.1 | 14 | 1882 | ||
NM_001323412.1 | NP_001310341.1 | 13 | 1673 | |||
NM_001323413.1 | NP_001310342.1 | 14 | 2039 | |||
NM_001323414.1 | NP_001310343.1 | 14 | 1908 | |||
NM_001323415.1 | NP_001310344.1 | 14 | 2033 | |||
NM_001323416.1 | NP_001310345.1 | 13 | 1824 | |||
NM_001323417.1 | NP_001310346.1 | 13 | 1783 | |||
NM_001323418.1 | NP_001310347.1 | 13 | 1818 | |||
NM_001323419.1 | NP_001310348.1 | 11 | 1418 | |||
NM_002860.3 | NP_002851.2 | 14 | 1888 | |||
GenBank mRNA | AK225455.1 | - | 13 | 1458 | ||
AK225465.1 | - | 13 | 1458 | |||
AK226032.1 | - | 13 | 1458 | |||
AK295487.1 | - | 13 | 1539 | |||
AK299557.1 | - | 11 | 1316 | |||
AK312271.1 | - | 14 | 1790 | |||
BC106054.1 | - | 14 | 1723 | 6156087 | ||
BC117240.1 | - | 14 | 1829 | 40125791 | ||
BC117242.1 | - | 14 | 1829 | 40125793 | ||
BC143930.1 | - | 14 | 1823 | 9052448 | ||
U68758.1 | - | 14 | 1697 | |||
U76542.1 | - | 14 | 1703 | |||
X94453.1 | - | 14 | 1784 |