Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENTPD1 SIRNA Assays ›
Gene Symbol:
ENTPD1
Gene Name:
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases:
ATPDase, CD39, NTPDase-1, SPG64
Chromosome Location:
Chr.10: 95710901 - 95877266 on Build GRCh38
UniGene ID:
Hs.576612
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534127
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENTPD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098175.1
NP_001091645.1
6
815
NM_001164178.1
NP_001157650.1
6
715
NM_001164179.1
NP_001157651.1
6
940
NM_001164181.1
NP_001157653.1
5
587
NM_001164182.1
NP_001157654.1
5
789
NM_001164183.1
NP_001157655.1
4
671
NM_001312654.1
NP_001299583.1
6
761
NM_001320916.1
NP_001307845.1
6
715
NM_001776.5
NP_001767.3
6
940
XM_011540370.2
XP_011538672.2
5
675
XM_011540371.2
XP_011538673.1
7
951
XM_011540372.2
XP_011538674.2
5
722
XM_011540373.2
XP_011538675.2
5
718
XM_011540374.2
XP_011538676.2
5
690
XM_011540376.2
XP_011538678.2
5
741
XM_011540377.2
XP_011538679.1
6
822
XM_017016958.1
XP_016872447.1
6
1082
XM_017016959.1
XP_016872448.1
4
405
XM_017016960.1
XP_016872449.1
5
561
XM_017016961.1
XP_016872450.1
5
546
XM_017016962.1
XP_016872451.1
5
727
XM_017016963.1
XP_016872452.1
6
707
XM_017016964.1
XP_016872453.1
6
1082
GenBank mRNA
AJ133133.1
-
6
820
AJ133134.1
-
6
820
AK298648.1
-
6
686
AK301459.1
-
6
704
AK304018.1
-
6
701
AK304836.1
-
5
543
AK316009.1
-
5
492
BC047664.1
-
4
408
5762493
S73813.1
-
6
683
U87967.1
-
6
646
Back To Top
More Information
Panther Classification:
Molecular Function -
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
blood coagulation
protein binding
ATP binding
hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline