Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPIN1 SIRNA Assays ›
Gene Symbol:
LPIN1
Gene Name:
lipin 1
Gene Aliases:
PAP1
Chromosome Location:
Chr.2: 11677544 - 11827409 on Build GRCh38
UniGene ID:
Hs.467740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534136
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LPIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261427.1
NP_001248356.1
6
976
NM_001261428.1
NP_001248357.1
6
897
NM_001261429.1
NP_001248358.1
6
1079
NM_145693.2
NP_663731.1
5
766
XM_006711869.2
XP_006711932.1
6
932
XM_006711870.3
XP_006711933.1
6
1063
XM_006711871.2
XP_006711934.1
5
766
XM_006711872.2
XP_006711935.1
5
990
XM_006711874.2
XP_006711937.1
6
2594
XM_011510333.2
XP_011508635.1
6
932
XM_011510334.2
XP_011508636.1
6
959
XM_011510335.2
XP_011508637.1
8
1422
XM_011510336.2
XP_011508638.1
7
1304
XM_017003623.1
XP_016859112.1
6
796
XM_017003624.1
XP_016859113.1
6
991
XM_017003625.1
XP_016859114.1
5
708
XM_017003626.1
XP_016859115.1
5
766
XM_017003627.1
XP_016859116.1
8
1422
XM_017003628.1
XP_016859117.1
8
1422
XM_017003629.1
XP_016859118.1
6
991
XM_017003630.1
XP_016859119.1
5
708
XM_017003631.1
XP_016859120.1
4
690
XM_017003632.1
XP_016859121.1
4
690
GenBank mRNA
AK290235.1
-
5
726
AK294742.1
-
6
881
AK294853.1
-
1
50
AK302922.1
-
6
955
AK310450.1
-
6
616
BC030537.1
-
5
759
3906281
D80010.1
-
5
709
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
triglyceride mobilization
phosphatidylethanolamine biosynthetic process
phosphatidylcholine biosynthetic process
mitotic nuclear envelope disassembly
fatty acid catabolic process
dephosphorylation
triglyceride biosynthetic process
organ regeneration
cellular response to insulin stimulus
positive regulation of transcription from RNA polymerase II promoter
molecular_function
transcription coactivator activity
phosphatidate phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline