Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000834.3 | NP_000825.2 | 13 | 3910 | ||
XM_005253351.3 | XP_005253408.1 | 4 | 1637 | |||
XM_011520628.2 | XP_011518930.1 | 14 | 4703 | |||
XM_011520629.2 | XP_011518931.1 | 15 | 4510 | |||
XM_017019219.1 | XP_016874708.1 | 14 | 4258 | |||
GenBank mRNA | AB208850.1 | - | 10 | 2689 | ||
BC113618.1 | - | 13 | 3809 | 8322670 | ||
BC113620.1 | - | 13 | 3809 | 8322672 | ||
U11287.1 | - | 13 | 3941 | |||
U88963.1 | - | 13 | 3910 | |||
U90278.1 | - | 13 | 3940 |