Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015878.1 | NP_001015878.1 | 4 | 593 | ||
NM_001015879.1 | NP_001015879.1 | 4 | 452 | |||
NM_003160.2 | NP_003151.2 | 4 | 457 | |||
GenBank mRNA | AB017332.1 | - | 4 | 598 | ||
AF054621.1 | - | 4 | 598 | |||
AF059681.1 | - | 4 | 653 | |||
AK301886.1 | - | 3 | 1033 | |||
AY661554.1 | - | 4 | 452 | |||
AY714054.1 | - | 4 | 395 | |||
BC075064.2 | - | 3 | 346 | 30915373 | ||
EU668345.1 | - | 4 | 653 |