Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166247.1 | NP_001159719.1 | 8 | 1452 | ||
NM_021956.4 | NP_068775.1 | 8 | 1452 | |||
NM_175768.3 | NP_786944.1 | 8 | 1452 | |||
XM_005266945.2 | XP_005267002.1 | 9 | 1582 | |||
XM_005266946.3 | XP_005267003.1 | 8 | 1537 | |||
XM_011535777.2 | XP_011534079.1 | 8 | 1619 | |||
XM_011535778.2 | XP_011534080.1 | 8 | 1619 | |||
XM_017010781.1 | XP_016866270.1 | 8 | 1619 | |||
XM_017010782.1 | XP_016866271.1 | 8 | 1619 | |||
GenBank mRNA | AJ252246.1 | - | 8 | 1159 | ||
AJ301610.1 | - | 8 | 1159 | |||
AK289542.1 | - | 8 | 1386 | |||
AK291965.1 | - | 8 | 1408 | |||
AK303721.1 | - | 8 | 1162 | |||
BC063814.1 | - | 8 | 1362 | 6189201 | ||
HM149335.1 | - | 8 | 1159 | |||
HM149336.1 | - | 8 | 1159 | |||
HM149337.1 | - | 8 | 1159 | |||
HM149338.1 | - | 8 | 1159 | |||
HM149339.1 | - | 8 | 1159 | |||
U16126.1 | - | 8 | 1159 |