Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 50 | 4048 | ||
GenBank mRNA | AK226074.1 | - | 50 | 3699 | ||
AL833478.1 | - | 7 | 4356 | |||
BC042586.1 | - | 50 | 3695 | 4803351 | ||
BC054498.1 | - | 50 | 3677 | 6474549 | ||
KU177988.1 | - | 50 | 3577 | |||
X55525.1 | - | 7 | 702 | |||
Z74616.1 | - | 50 | 3716 |