Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000288.3 | NP_000279.1 | 10 | 1055 | ||
XM_005267019.4 | XP_005267076.1 | 10 | 1335 | |||
XM_006715502.1 | XP_006715565.1 | 7 | 1105 | |||
XM_017010934.1 | XP_016866423.1 | 7 | 1122 | |||
GenBank mRNA | AL583681.3 | - | 1 | 52 | ||
BC006268.1 | - | 10 | 997 | 3948721 | ||
BC031606.1 | - | 11 | 1147 | 5176358 | ||
CR542054.1 | - | 10 | 953 | |||
CR542076.1 | - | 10 | 953 | |||
U76560.1 | - | 10 | 1001 | |||
U88871.1 | - | 10 | 1034 |