Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000288.3 | NP_000279.1 | 2 | 253 | ||
XM_005267019.4 | XP_005267076.1 | 2 | 533 | |||
XM_006715502.1 | XP_006715565.1 | 2 | 597 | |||
XM_017010934.1 | XP_016866423.1 | 2 | 597 | |||
GenBank mRNA | BC006268.1 | - | 2 | 195 | 3948721 | |
BC031606.1 | - | 2 | 237 | 5176358 | ||
BP246238.1 | - | 2 | 253 | |||
CR542054.1 | - | 2 | 151 | |||
CR542076.1 | - | 2 | 151 | |||
U76560.1 | - | 2 | 199 | |||
U88871.1 | - | 2 | 232 |