Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000550.2 | NP_000541.1 | 7 | 1454 | ||
GenBank mRNA | AK297887.1 | - | 5 | 696 | ||
BC052608.1 | - | 6 | 1373 | 6258285 | ||
CR407683.1 | - | 6 | 1276 | |||
X51420.1 | - | 7 | 1405 | |||
X51455.1 | - | 5 | 627 |