Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534256
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
4
1077
NM_024790.6
NP_079066.5
6
924
XM_005251305.3
XP_005251362.2
9
1408
XM_006716474.2
XP_006716537.2
9
1408
XM_006716477.2
XP_006716540.2
9
1408
XM_011517598.1
XP_011515900.1
9
1408
XM_011517599.1
XP_011515901.1
8
1384
XM_011517600.1
XP_011515902.1
8
1384
XM_011517601.1
XP_011515903.1
7
1303
XM_011517602.1
XP_011515904.1
7
1303
XM_011517603.1
XP_011515905.1
8
1091
XM_011517607.1
XP_011515909.1
7
971
XM_011517608.1
XP_011515910.1
6
968
XM_011517609.1
XP_011515911.1
5
1090
XM_017013847.1
XP_016869336.1
9
1408
XM_017013848.1
XP_016869337.1
8
1384
XM_017013849.1
XP_016869338.1
7
1303
XM_017013850.1
XP_016869339.1
7
1303
XM_017013851.1
XP_016869340.1
6
968
XM_017013852.1
XP_016869341.1
7
1303
XM_017013853.1
XP_016869342.1
6
968
XM_017013854.1
XP_016869343.1
8
1384
XM_017013855.1
XP_016869344.1
7
1303
XM_017013856.1
XP_016869345.1
6
968
XM_017013857.1
XP_016869346.1
2
477
XM_017013858.1
XP_016869347.1
2
477
GenBank mRNA
AJ583433.1
-
4
1077
AK096205.1
-
7
978
AK127108.1
-
8
969
AM156947.1
-
6
925
BX649147.1
-
6
925
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline