Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017653.3 | NP_060123.3 | 15 | 2054 | ||
XM_006722488.2 | XP_006722551.1 | 15 | 2049 | |||
XM_006722490.2 | XP_006722553.1 | 15 | 2049 | |||
XM_011526036.1 | XP_011524338.1 | 15 | 2049 | |||
XM_011526037.1 | XP_011524339.1 | 15 | 2051 | |||
XM_011526038.1 | XP_011524340.1 | 15 | 2046 | |||
XM_011526039.1 | XP_011524341.1 | 15 | 2049 | |||
XM_011526040.1 | XP_011524342.1 | 16 | 2050 | |||
XM_011526042.1 | XP_011524344.1 | 15 | 2049 | |||
XM_017025795.1 | XP_016881284.1 | 15 | 2048 | |||
XM_017025796.1 | XP_016881285.1 | 16 | 2060 | |||
XM_017025797.1 | XP_016881286.1 | 15 | 1987 | |||
XM_017025798.1 | XP_016881287.1 | 15 | 2046 | |||
GenBank mRNA | AK000078.1 | - | 11 | 1367 | ||
AK074611.1 | - | 15 | 2049 | |||
AK091256.1 | - | 16 | 2023 | |||
AK225205.1 | - | 11 | 1367 | |||
AK291303.1 | - | 15 | 2019 | |||
AK296579.1 | - | 10 | 1441 | |||
AK315091.1 | - | 16 | 1995 | |||
AL390156.1 | - | 4 | 356 | |||
BC001252.1 | - | 12 | 1453 | |||
BC064394.1 | - | 15 | 2036 | 5752466 |