Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017653.3 | NP_060123.3 | 14, 15 | 1937 | ||
XM_006722488.2 | XP_006722551.1 | 14, 15 | 1932 | |||
XM_006722490.2 | XP_006722553.1 | 14, 15 | 1932 | |||
XM_011526036.1 | XP_011524338.1 | 14, 15 | 1932 | |||
XM_011526037.1 | XP_011524339.1 | 14, 15 | 1934 | |||
XM_011526038.1 | XP_011524340.1 | 14, 15 | 1929 | |||
XM_011526039.1 | XP_011524341.1 | 14, 15 | 1932 | |||
XM_011526040.1 | XP_011524342.1 | 15, 16 | 1933 | |||
XM_011526042.1 | XP_011524344.1 | 14, 15 | 1932 | |||
XM_017025795.1 | XP_016881284.1 | 14, 15 | 1931 | |||
XM_017025796.1 | XP_016881285.1 | 15, 16 | 1943 | |||
XM_017025797.1 | XP_016881286.1 | 14, 15 | 1870 | |||
XM_017025798.1 | XP_016881287.1 | 14, 15 | 1929 | |||
GenBank mRNA | AK000078.1 | - | 10, 11 | 1250 | ||
AK074611.1 | - | 14, 15 | 1932 | |||
AK091256.1 | - | 15, 16 | 1906 | |||
AK225205.1 | - | 10, 11 | 1250 | |||
AK291303.1 | - | 14, 15 | 1902 | |||
AK296579.1 | - | 9, 10 | 1324 | |||
AK315091.1 | - | 15, 16 | 1878 | |||
AL390156.1 | - | 3, 4 | 239 | |||
BC001252.1 | - | 11, 12 | 1336 | |||
BC064394.1 | - | 14, 15 | 1919 | 5752466 |