Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271938.1 | NP_001258867.1 | 39 | 7626 | ||
NM_001410.2 | NP_001401.2 | 38 | 7425 | |||
GenBank mRNA | AB011541.2 | - | 38 | 7425 | ||
AY280362.1 | - | 33 | 5748 | |||
BC033228.1 | - | 2 | 250 | |||
BC153880.1 | - | 38 | 7425 | 8860114 |