Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 11 | 3814 | ||
NM_001282543.1 | NP_001269472.1 | 10 | 3757 | |||
NM_001282545.1 | NP_001269474.1 | 7 | 2461 | |||
NM_001282548.1 | NP_001269477.1 | 6 | 2404 | |||
NM_001282549.1 | NP_001269478.1 | 5 | 2275 | |||
NR_104212.1 | 10 | 3665 | ||||
NR_104215.1 | 9 | 3608 | ||||
NR_104216.1 | 10 | 2864 | ||||
XM_017004613.1 | XP_016860102.1 | 12 | 3862 |