Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005996.3 | NP_005987.3 | 3, 4 | 1776 | ||
NM_016569.3 | NP_057653.3 | 4, 5 | 1836 | |||
GenBank mRNA | AF002228.1 | - | 3, 4 | 812 | ||
AF170708.2 | - | 3, 4 | 1072 | |||
AF216750.1 | - | 4, 5 | 988 | |||
AK054604.1 | - | 3, 4 | 1779 | |||
AY034105.1 | - | 2, 3 | 258 | |||
BC025258.1 | - | 4, 5 | 1262 | 5458526 |