Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 4, 5 | 491 | ||
GenBank mRNA | AK300070.1 | - | 4, 5 | 380 | ||
AK314340.1 | - | 4, 5 | 473 | |||
BC015045.2 | - | 4, 5 | 416 | 3922902 | ||
BI910081.1 | - | 4, 5 | 393 | |||
BT009852.1 | - | 4, 5 | 375 | |||
KU178089.1 | - | 4, 5 | 375 | |||
KU178090.1 | - | 4, 5 | 375 | |||
M60091.1 | - | 4, 5 | 403 |