Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 5, 6 | 624 | ||
NM_001258332.1 | NP_001245261.1 | 3, 4 | 499 | |||
GenBank mRNA | AK096137.1 | - | 4, 5 | 663 | ||
AK293212.1 | - | 3, 4 | 782 | |||
AK300070.1 | - | 5, 6 | 513 | |||
AK303279.1 | - | 3, 4 | 452 | |||
AK314340.1 | - | 5, 6 | 606 | |||
BC015045.2 | - | 5, 6 | 549 | 3922902 | ||
BI910081.1 | - | 5, 6 | 526 | |||
BT009852.1 | - | 5, 6 | 508 | |||
KU178089.1 | - | 5, 6 | 508 | |||
M60091.1 | - | 5, 6 | 536 |