Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000155.3 | NP_000146.2 | 1 | 189 | ||
NM_001258332.1 | NP_001245261.1 | 1 | 189 | |||
GenBank mRNA | AK096026.1 | - | 1 | 103 | ||
AK096137.1 | - | 1 | 103 | |||
AK293212.1 | - | 1 | 132 | |||
AK303279.1 | - | 1 | 142 | |||
AK310049.1 | - | 1 | 103 | |||
AK314340.1 | - | 1 | 171 | |||
BC015045.2 | - | 1 | 114 | 3922902 | ||
BI910081.1 | - | 1 | 140 | |||
BT009852.1 | - | 1 | 73 | |||
DA011092.1 | - | 1 | 189 | |||
KU178089.1 | - | 1 | 73 | |||
KU178090.1 | - | 1 | 73 | |||
M60091.1 | - | 1 | 101 |