Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001609.3 | NP_001600.1 | 10, 11 | 1349 | ||
XM_017016194.1 | XP_016871683.1 | 9, 10 | 1190 | |||
GenBank mRNA | AK298638.1 | - | 9, 10 | 1178 | ||
AK314241.1 | - | 10, 11 | 1245 | |||
AL831821.2 | - | 10, 11 | 1218 | |||
BC013756.1 | - | 10, 11 | 1244 | 4706281 | ||
KU177879.1 | - | 10, 11 | 1232 | |||
KU177880.1 | - | 11, 12 | 1540 | |||
U12778.1 | - | 10, 11 | 1247 |