Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000433.3 | NP_000424.2 | 9 | 1155 | ||
NM_001127651.2 | NP_001121123.1 | 10 | 992 | |||
NM_001190789.1 | NP_001177718.1 | 7 | 912 | |||
NM_001190794.1 | NP_001177723.1 | 8 | 1020 | |||
XM_005245207.1 | XP_005245264.1 | 8 | 976 | |||
XM_011509580.1 | XP_011507882.1 | 10 | 993 | |||
XM_011509581.1 | XP_011507883.1 | 10 | 980 | |||
GenBank mRNA | AB209647.1 | - | 7 | 4425 | ||
AF527950.1 | - | 9 | 902 | |||
AK296672.1 | - | 7 | 768 | |||
AK298713.1 | - | 8 | 929 | |||
AK312666.1 | - | 9 | 1065 | |||
BC001606.1 | - | 9 | 1132 | 3542841 | ||
BT007439.1 | - | 9 | 880 | |||
KU178216.1 | - | 9 | 880 | |||
M32011.1 | - | 10 | 947 |