Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000158.3 | NP_000149.3 | 9 | 1436 | ||
GenBank mRNA | AB209731.1 | - | 9 | 1326 | ||
AK125918.1 | - | 9 | 1799 | |||
AK297939.1 | - | 7 | 869 | |||
AK300620.1 | - | 9 | 1083 | |||
AK312245.1 | - | 9 | 1285 | |||
BC012098.1 | - | 9 | 1365 | 4574938 | ||
L07956.1 | - | 9 | 1243 |