Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719050.3 | XP_006719113.1 | 26 | 3117 | ||
XM_011520592.2 | XP_011518894.1 | 26 | 3113 | |||
XM_017018918.1 | XP_016874407.1 | 27 | 3295 | |||
XM_017018919.1 | XP_016874408.1 | 28 | 3358 | |||
XM_017018920.1 | XP_016874409.1 | 27 | 3257 | |||
XM_017018922.1 | XP_016874411.1 | 27 | 3088 | |||
XM_017018923.1 | XP_016874412.1 | 27 | 2981 | |||
XM_017018924.1 | XP_016874413.1 | 27 | 3014 | |||
XM_017018925.1 | XP_016874414.1 | 27 | 3086 | |||
XM_017018926.1 | XP_016874415.1 | 27 | 3155 | |||
XM_017018927.1 | XP_016874416.1 | 26 | 3146 | |||
XM_017018928.1 | XP_016874417.1 | 27 | 3300 | |||
XM_017018929.1 | XP_016874418.1 | 27 | 3262 | |||
XM_017018931.1 | XP_016874420.1 | 26 | 2913 | |||
XM_017018933.1 | XP_016874422.1 | 27 | 3382 | |||
XM_017018934.1 | XP_016874423.1 | 25 | 2834 | |||
XM_017018935.1 | XP_016874424.1 | 23 | 2318 | |||
XM_017018936.1 | XP_016874425.1 | 23 | 3313 | |||
XM_017018937.1 | XP_016874426.1 | 23 | 3318 | |||
GenBank mRNA | U75968.1 | - | 27 | 3127 |
Gene Symbol: DDX12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065494.1 | - | 15 | 2844 |