Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005253331.3 | XP_005253388.1 | 1 | 234 | ||
XM_005253333.3 | XP_005253390.1 | 1 | 234 | |||
XM_006719050.3 | XP_006719113.1 | 1 | 234 | |||
XM_011520592.2 | XP_011518894.1 | 1 | 235 | |||
XM_011520595.2 | XP_011518897.1 | 1 | 234 | |||
XM_011520597.2 | XP_011518899.1 | 1 | 234 | |||
XM_017018918.1 | XP_016874407.1 | 1 | 237 | |||
XM_017018919.1 | XP_016874408.1 | 2 | 300 | |||
XM_017018920.1 | XP_016874409.1 | 1 | 238 | |||
XM_017018921.1 | XP_016874410.1 | 1 | 237 | |||
XM_017018926.1 | XP_016874415.1 | 1 | 238 | |||
XM_017018927.1 | XP_016874416.1 | 1 | 238 | |||
XM_017018928.1 | XP_016874417.1 | 1 | 237 | |||
XM_017018929.1 | XP_016874418.1 | 1 | 238 | |||
XM_017018930.1 | XP_016874419.1 | 1 | 236 | |||
XM_017018938.1 | XP_016874427.1 | 1 | 236 | |||
XM_017018939.1 | XP_016874428.1 | 1 | 236 |