Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005213.3 | NP_005204.1 | 1, 2 | 188 | ||
GenBank mRNA | AK291308.1 | - | 1, 2 | 104 | ||
AV716538.1 | - | 1, 2 | 202 | |||
AV717631.1 | - | 1, 2 | 97 | |||
BC010379.1 | - | 1, 2 | 108 | 4063525 | ||
CR456914.1 | - | 1, 2 | 59 | |||
X05978.1 | - | 1, 2 | 104 |