Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000336.2 | NP_000327.2 | 3 | 719 | ||
XM_011545913.2 | XP_011544215.1 | 4 | 711 | |||
XM_011545914.1 | XP_011544216.1 | 3 | 567 | |||
XM_017023525.1 | XP_016879014.1 | 4 | 729 | |||
XM_017023526.1 | XP_016879015.1 | 4 | 729 | |||
GenBank mRNA | AK313192.1 | - | 3 | 644 | ||
BC036352.1 | - | 3 | 678 | 5178288 | ||
DQ402523.1 | - | 2 | 543 | |||
L36593.1 | - | 2 | 543 | |||
X87159.1 | - | 3 | 670 |