Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCNKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000085.4 | NP_000076.2 | 10 | 1070 | ||
NM_001165945.2 | NP_001159417.2 | 3 | 621 | |||
GenBank mRNA | AK098217.1 | - | 3 | 594 | ||
AK292335.1 | - | 10 | 1054 | |||
DA628551.1 | - | 6 | 517 | |||
S80315.1 | - | 10 | 942 | |||
Z30644.1 | - | 10 | 952 |