Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCNKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000085.4 | NP_000076.2 | 18 | 2072 | ||
NM_001165945.2 | NP_001159417.2 | 11 | 1620 | |||
GenBank mRNA | AK098217.1 | - | 11 | 1593 | ||
AK292335.1 | - | 18 | 2056 | |||
S80315.1 | - | 18 | 1944 | |||
Z30644.1 | - | 18 | 1954 |