Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017979.2 | NP_001017979.1 | 3 | 403 | ||
NM_001159601.1 | NP_001153073.1 | 3 | 403 | |||
NM_004249.3 | NP_004240.2 | 3 | 403 | |||
XM_005248218.4 | XP_005248275.1 | 3 | 403 | |||
XM_011513911.2 | XP_011512213.1 | 3 | 403 | |||
XM_011513912.2 | XP_011512214.1 | 3 | 1404 | |||
XM_017008850.1 | XP_016864339.1 | 3 | 403 | |||
GenBank mRNA | AF498955.1 | - | 3 | 188 | ||
BC018067.1 | - | 3 | 369 | |||
BC035054.2 | - | 3 | 376 | 5263482 | ||
CX165950.1 | - | 3 | 223 | |||
DA369194.1 | - | 3 | 403 | |||
X94703.1 | - | 3 | 188 |