Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139517.1 | NP_001132989.1 | 1 | 393 | ||
NM_001139518.1 | NP_001132990.1 | 2 | 114 | |||
NM_001316362.1 | NP_001303291.1 | 2 | 322 | |||
NM_003690.4 | NP_003681.1 | 2 | 313 | |||
XM_011512063.1 | XP_011510365.1 | 2 | 195 | |||
XM_011512066.1 | XP_011510368.1 | 2 | 359 | |||
XM_017005159.1 | XP_016860648.1 | 2 | 195 | |||
GenBank mRNA | AF072860.1 | - | 2 | 208 | ||
AF083033.1 | - | 2 | 204 | |||
AK223107.1 | - | 2 | 212 | |||
AK290601.1 | - | 2 | 239 | |||
AK296019.1 | - | 2 | 233 | |||
AL136615.1 | - | 2 | 237 | |||
AL833867.1 | - | 1 | 393 | |||
AY251164.1 | - | 2 | 131 | |||
BC009470.1 | - | 2 | 219 | 3503747 | ||
BG827125.1 | - | 2 | 314 | |||
BT007243.1 | - | 2 | 112 | |||
CR533525.1 | - | 2 | 112 | |||
HY049565.1 | - | 2 | 322 |